1-Ethylimidazole for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: N-Ethyl-1H-imidazole, 1-Ethyl-1H-imidazole, N-Ethylimidazole

M-Clarity™ quality level = MQ200

8.43830.0010 8.43830.0100
8.43830.0010EA 60.2 EUR
8.43830.0010 8.43830.0100
1-Ethylimidazole for synthesis, Sigma-Aldrich®
1-Ethylimidazole
Formula: C₅H₈N₂
MW: 96.13 g/mol
Density: 0.995 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00055389
CAS Number: 7098-07-9
EINECS: 230-403-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR