1-Benzylimidazole 99%
Supplier: Thermo Fisher Scientific
Synonyms:
1-Benzyl-1H-imidazole
152250050
152250250
152251000
ACRO152250050EA
45.2
EUR
ACRO152250050
ACRO152250250
ACRO152251000
1-Benzylimidazole 99%
1-Benzylimidazole
Formula:
C₁₀H₁₀N₂ Boiling Pt: 310 °C (760 mmHg) Melting Pt: 68…71 °C Density: 1,22 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005296 CAS Number: 4238-71-5 EINECS: 224-200-4 |
Specification Test Results
Appearance | Brown Crystals |
Infrared spectrum | Conforms |
Melting point | 68 to 71 °C |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...