Indole-4-carbonitrile
Supplier: Cayman Chemical
|
Warning
|
Synonyms:
4-Cyano-1H-indole, 1H-Indole-4-carbonitrile, 4-Cyanoindole
10007084-1
10007084-10
10007084-5
10007084-500
CAYM10007084-1EA
121
EUR
CAYM10007084-1
CAYM10007084-10
CAYM10007084-5
CAYM10007084-500
Indole-4-carbonitrile
Indole-4-carbonitrile
Formula:
C₉H₆N₂ MW: 142,16 g/mol |
MDL Number:
MFCD00152045 CAS Number: 16136-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...