4-Nitroindole ≥97%
Supplier: Thermo Scientific
Synonyms:
4-Nitro-1H-indole
L09177.03
L09177.MC
L09177.ME
L09177.03EA
189
EUR
L09177.03
L09177.MC
L09177.ME
4-Nitroindole ≥97%
4-Nitroindole
Purity: 97%
CAS No.: 4769-97-5
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Melting Point: 202-207°
Beilstein Reference: 136022
Formula:
C₈H₆N₂O₂ MW: 162,15 g/mol Melting Pt: 202…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010056 CAS Number: 4769-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...