4-Methoxyindole ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Methoxy-1H-indole, Indol-4-yl methyl ether
L19069.03
L19069.03EA
139
EUR
L19069.03
4-Methoxyindole ≥99%
4-Methoxyindole
Purity: 99%
CAS No.: 4837-90-5
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Melting Point: 69-71°
Boiling Point: 181-183°/24mm
Beilstein Reference: 122456
MDL No.: MFCD00009737
Formula:
C₉H₉NO MW: 147,18 g/mol Boiling Pt: 181…183 °C (24 mmHg) Melting Pt: 69…71 °C |
MDL Number:
MFCD00009737 CAS Number: 4837-90-5 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...