5-Methoxyindole ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Methoxy-1H-indole, Indol-5-yl methyl ether

B25290.03 B25290.06 B25290.14
B25290.03EA 37.8 EUR
B25290.03 B25290.06 B25290.14
5-Methoxyindole ≥99%
5-Methoxyindole

Purity: 99% CAS No.: 1006-94-6 Molecular Formula: C9H9NO Formula Weight: 147.18 Storage Temperature: 2°C to 8°C Melting Point: 52-55° Boiling Point: 176-178°/17mm Beilstein Reference: 116722 MDL No.: MFCD00005674

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 176…178 °C (17 mmHg)
Melting Pt: 52…55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005674
CAS Number: 1006-94-6
EINECS: 213-745-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR