6-Methoxyindole ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether

L19074.03 L19074.MD
L19074.03EA 144 EUR
L19074.03 L19074.MD
6-Methoxyindole ≥98%
6-Methoxyindole

Purity: 98+% CAS No.: 3189-13-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 90-94° Boiling Point: 105°/0.2mm MDL No.: MFCD00022780

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 105 °C (0.2 mmHg)
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00022780
CAS Number: 3189-13-7
EINECS: 221-689-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR