Indole 99+%

Supplier: Thermo Scientific
Danger

Synonyms: 1H-Indole

122150100 122150250 122151000 122155000 122150025
ACRO122150100EA 27.8 EUR
ACRO122150100 ACRO122150250 ACRO122151000 ACRO122155000 ACRO122150025
Indole 99+%
Indole
Formula: C₈H₇N
MW: 117,15 g/mol
Boiling Pt: 253…254 °C (760 mmHg)
Melting Pt: 51…53 °C
Density: 1,22 g/cm³
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,04985

Order Now

Specification Test Results

Appearance White to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 51 to 53 °C
GC ≥99.0 %
UV at 195 nm A: ≤1.0
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR