Cerium trichloride, anhydrous 99.9%, pure

Supplier: Thermo Scientific
Danger

Synonyms: Cerous chloride, Cerium(III) chloride, Cerium (III) chloride

369750050 369750250 369751000
ACRO369750050EA 107 EUR
ACRO369750050 ACRO369750250 ACRO369751000
Cerium trichloride, anhydrous 99.9%, pure
Cerium trichloride
Formula: CeCl₃
MW: 246,47 g/mol
Boiling Pt: 1727 °C (760 mmHg)
Melting Pt: 848 °C
Density: 3,193 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010929
CAS Number: 7790-86-5
EINECS: 232-227-8
UN: 1759
ADR: 8,II
Merck Index: 13,02008

Order Now

Specification Test Results

Appearance White to light yellow Powder or granules
Total conc. of imp.as REO ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR