Dysprosium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
|
Warning
|
41409.09
41409.18
41409.09EA
172
EUR
41409.09
41409.18
Dysprosium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Dysprosium trifluoride
Purity: 99.9%
CAS No.: 13569-80-7
Molecular Formula: DyF3
Formula Weight: 219.50
Physical Form: 12mm & down Pieces
Density: 7.5
Appearance: White
Odour: Odourless
Melting Point: 1360°
Boiling Point: 2200°
MDL No.: MFCD00016071
For vacuum deposition
Formula:
DyF₃ MW: 219,5 g/mol Boiling Pt: 2200 °C Melting Pt: 1360 °C Density: 7,5 Storage Temperature: Ambient |
MDL Number:
MFCD00016071 CAS Number: 13569-80-7 EINECS: 236-992-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...