Thulium(III) iodide ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Fisher Scientific
Warning

42094.03 42094.06
42094.03EA 152 EUR
42094.03 42094.06
Thulium(III) iodide ≥99.9% (REO, rare earth oxide basis)
Thulium (III) iodide

Purity: 99.9% CAS No.: 13813-43-9 Molecular Formula: TmI3 Formula Weight: 549.65 Physical Form: -20 Mesh Melting Point: 1015° Boiling Point: 1260° MDL No.: MFCD00050141

Formula: I₃Tm
MW: 549.65 g/mol
Boiling Pt: 1260 °C
Storage Temperature: Ambient
MDL Number: MFCD00050141
CAS Number: 13813-43-9
EINECS: 237-472-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR