Cerium(IV) oxide ≥99.5%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Ceric oxide, Cerium dioxide
12925.22
12925.36
12925.A3
12925.22EA
60.1
EUR
12925.22
12925.36
12925.A3
Cerium(IV) oxide ≥99.5%
Cerium (IV) oxide
Purity: 99.5%
CAS No.: 1306-38-3
Molecular Formula: CeO2
Formula Weight: 172.12
Physical Form: Powder
Density: 7.132
Appearance: Pale yellow
Odour: Odourless
Melting Point: 2600°
Merck Reference: 14,1989
MDL No.: MFCD00010933
Formula:
CeO₂ MW: 172.11 g/mol Boiling Pt: 3468 Melting Pt: 2600 °C Density: 7.132 Storage Temperature: Ambient |
MDL Number:
MFCD00010933 CAS Number: 1306-38-3 EINECS: 215-150-4 Merck Index: 12,02035 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...