Europium(III) oxide 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
193020010
193020050
193020250
ACRO193020010EA
48
EUR
ACRO193020010
ACRO193020050
ACRO193020250
Europium(III) oxide 99.99% (trace metals basis)
Europium (III) oxide
Formula:
Eu₂O₃ Melting Pt: 2325 °C Density: 7,42 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010997 CAS Number: 1308-96-9 EINECS: 215-165-6 Merck Index: 14,03904 |
Specification Test Results
Appearance | White to light pink Powder |
Total conc. of imp.as REO | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...