Digadolinium trioxide ≥99.99% (REO, rare earth oxide basis), nanopowder
Supplier: Thermo Fisher Scientific
|
Warning
|
44020.14
44020.22
44020.36
44020.14EA
190
EUR
44020.14
44020.22
44020.36
Digadolinium trioxide ≥99.99% (REO, rare earth oxide basis), nanopowder
Digadolinium trioxide
Purity: 99.99+%
CAS No.: 12064-62-9
Molecular Formula: Gd2O3
Formula Weight: 362.50
Physical Form: 20-40nm Powder
Density: 7.407
Melting Point: 2330°
Merck Reference: 14,4326
MDL No.: MFCD00011026
Formula:
Gd₂O₃ Melting Pt: 2330 °C Density: 7,407 Storage Temperature: Ambient |
MDL Number:
MFCD00011026 CAS Number: 12064-62-9 EINECS: 235-060-9 Merck Index: 14,4326 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...