Safety shoes, lace-up, Basic, jo_Colour Yellow, Jori
Supplier: ELTEN
12931/36 12931/37 12931/38 12931/39 12931/40 12931/41 12931/42 12931/43 12931/44 12931/45 12931/46 12931/47 12931/48
ELTN12931/36EA 49.9 EUR
ELTN12931/36 ELTN12931/37 ELTN12931/38 ELTN12931/39 ELTN12931/40 ELTN12931/41 ELTN12931/42 ELTN12931/43 ELTN12931/44 ELTN12931/45 ELTN12931/46 ELTN12931/47 ELTN12931/48
Safety shoes, lace-up, Basic, jo_Colour Yellow, Jori
Shoes and Boots
Safety shoes made from quick drying, tear resistant textile upper combined with reflective materials, skin friendly textile lining, a full padded bellows tongue and collar padding plus a dual-density PU/PU (New Basic) sole (oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole
  • PU scuff cap protects the upper material
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/yellow
Certifications: EN ISO 20345; S1P, SRC