2-Naphthyl boronic acid 97+%
Supplier: Thermo Fisher Scientific
|
Warning
|
359620010
359620050
ACRO359620010EA
38.4
EUR
ACRO359620010
ACRO359620050
2-Naphthyl boronic acid 97+%
2-Naphthyl boronic acid
Formula:
C₁₀H₉BO₂ Melting Pt: 269…275 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00236051 CAS Number: 32316-92-0 |
Specification Test Results
Appearance | White to off-white Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...