2-Naphthyl boronic acid ≥97%

Supplier: Thermo Scientific
Warning

B24157.03 B24157.06 B24157.14
B24157.03EA 45 EUR
B24157.03 B24157.06 B24157.14
2-Naphthyl boronic acid ≥97%
2-Naphthyl boronic acid

Purity: 97% CAS No.: 32316-92-0 Molecular Formula: C10H9BO2 Formula Weight: 171.99 Storage Temperature: 2°C to 8°C Melting Point: ca 267° Beilstein Reference: 2936449 MDL No.: MFCD00236051

Formula: C₁₀H₉BO₂
Melting Pt: ∼ 267 °C
MDL Number: MFCD00236051
CAS Number: 32316-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR