Gold(I) chloride 99+%

Supplier: Thermo Fisher Scientific

388330010 388330050
ACRO388330010EA 340 EUR
ACRO388330010 ACRO388330050
Gold(I) chloride 99+%
Gold (I) chloride
Formula: AuCl
Melting Pt: 289 °C
Density: 7,57 g/cm³ (20 °C)
MDL Number: MFCD00046175
CAS Number: 10294-29-8
EINECS: 233-655-8
UN: 3260
ADR: 8,II
Merck Index: 12,04528

Order Now

Specification Test Results

Appearance Yellow-green Powder
Elemental Analysis ≥99.0 % (ex Au)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR