Gold(I) chloride ≥99.99% (metals basis) Au ≥ 84.2%, Premion®

Supplier: Thermo Fisher Scientific

40432.01 40432.03 40432.06
40432.01EA 110 EUR
40432.01 40432.03 40432.06
Gold(I) chloride ≥99.99% (metals basis) Au ≥ 84.2%, Premion®
Gold (I) chloride

Purity: 99.99% CAS No.: 10294-29-8 Molecular Formula: AuCl Formula Weight: 232.42 Physical Form: Powder Density: 7.57 Melting Point: 289° dec. Merck Reference: 14,4515 UN No.: 3260 MDL No.: MFCD00046175

Formula: AuCl
Melting Pt: 289 °C (dec.)
Density: 7,57
MDL Number: MFCD00046175
CAS Number: 10294-29-8
EINECS: 233-655-8
UN: 3260
ADR: 8,III
Merck Index: 14,4515

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR