Silver chloride ≥95%

Supplier: Thermo Fisher Scientific
Warning

87342.14 87342.22
87342.14EA 153 EUR
87342.14 87342.22
Silver chloride ≥95%
Silver chloride

Purity: 95% CAS No.: 7783-90-6 Molecular Formula: AgCl Formula Weight: 143.32 Physical Form: Coarse Powder Density: 5.56 Refractive Index: 2.071 Sensitivity: Light Sensitive Appearance: White Odour: Odourless Melting Point: 455° Boiling Point: 1550° Solubility: Practically insoluble in water. Soluble in ammonia, solutions of alkali cyanides, thiosulfates, ammonium carbonates. Insoluble in alcohol, dilute acids Merck Reference: 14,8509 UN No.: 3077 MDL No.: MFCD00003399


Silver plating

Formula: AgCl
MW: 143.32 g/mol
Boiling Pt: 1550 °C
Melting Pt: 455
Density: 5.56
Storage Temperature: Ambient
MDL Number: MFCD00003399
CAS Number: 7783-90-6
EINECS: 232-033-3
ADR: 9 III
Merck Index: 13,08582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR