Zinc chloride
Supplier: Roth Carl
|
Danger
|
Synonyms:
Zinc dichloride, Dichlorozinc, Zinc (II) chloride
3533.1
3533.2
ROTH3533.1EA
0
EUR
ROTH3533.1
ROTH3533.2
Zinc chloride
Zinc chloride
Formula:
ZnCl₂ MW: 136.3 g/mol Boiling Pt: 732 Melting Pt: 283 Density: 2.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8 III Merck Index: 13,10185 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...