Cadmium(II) fluoride ≥99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Cadmium fluoride
B20103.14
B20103.22
B20103.14EA
0
EUR
B20103.14
B20103.22
Cadmium(II) fluoride ≥99%
Cadmium (II) fluoride
Formula:
CdF₂ MW: 150,41 g/mol Boiling Pt: 1748 °C (1013 hPa) Melting Pt: 1065…1075 °C Density: 6,33 g/cm³ (20 °C) |
MDL Number:
MFCD00010919 CAS Number: 7790-79-6 EINECS: 232-222-0 UN: 2570 ADR: 6.1,II Merck Index: 12,01655 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...