Mercury(II) fluoride ≥95%

Supplier: Thermo Scientific
Danger

Synonyms: Mercury difluoride, Mercuric fluoride

11533.09 11533.18
11533.09EA 0 EUR
11533.09 11533.18
Mercury(II) fluoride ≥95%
Mercury (II) fluoride

Purity: 95% CAS No.: 7783-39-3 Molecular Formula: F2Hg Formula Weight: 238.61 Physical Form: -10 Mesh Crystalline Density: 8.95 Appearance: Yellow Odour: Odourless Melting Point: 645° dec. Merck Reference: 14,5878 UN No.: 2025 MDL No.: MFCD00016139

Formula: F₂Hg
MW: 238,59 g/mol
Boiling Pt: ∼650 °C (1013 hPa)
Melting Pt: 645 °C (dec.)
Density: 8,95
Storage Temperature: Ambient
MDL Number: MFCD00016139
CAS Number: 7783-39-3
EINECS: 231-994-6
UN: 2025
ADR: 6.1,II
Merck Index: 14,5878

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR