Zinc iodide 99.999% (trace metals basis), extra pure

Supplier: Thermo Fisher Scientific
Danger

212760050 212760250
ACRO212760050EA 72.3 EUR
ACRO212760050 ACRO212760250
Zinc iodide 99.999% (trace metals basis), extra pure
Zinc iodide
Formula: I₂Zn
Boiling Pt: 620 °C (1013 hPa)
Melting Pt: 446 °C
Density: 4,74 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS Number: 10139-47-6
EINECS: 233-396-0
UN: 3260
ADR: 8,II
Merck Index: 12,10269

Order Now

Specification Test Results

Appearance White to light yellow Powder
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR