Cadmium sulphide ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Cadmium (II) sulphide
45455.30
45455.18
45455.30EA
867
EUR
45455.30
45455.18
Cadmium sulphide ≥99.999% (metals basis)
Cadmium sulphide
Purity: 99.999%
CAS No.: 1306-23-6
Molecular Formula: CdS
Formula Weight: 144.46
Physical Form: 3-12mm Pieces
Density: 4.82
Appearance: Orange
Odour: Odourless
Melting Point: 980° subl.
Merck Reference: 14,1628
UN No.: 3077
MDL No.: MFCD00010922
Formula:
CdS Melting Pt: 980 °C (subl.) Density: 4,82 Storage Temperature: Ambient |
MDL Number:
MFCD00010922 CAS Number: 1306-23-6 EINECS: 215-147-8 UN: 3077 ADR: 9,III Merck Index: 14,1628 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...