Cadmium sulphide ≥98%

Supplier: Thermo Fisher Scientific
Danger

A14544.22 A14544.0E A14544.36
A14544.22EA 33.3 EUR
A14544.22 A14544.0E A14544.36
Cadmium sulphide ≥98%
Cadmium sulphide

Purity: 98% CAS No.: 1306-23-6 Molecular Formula: CdS Formula Weight: 144.46 Odour: Odourless Melting Point: 980° subl. UN No.: 3077 MDL No.: MFCD00010922

Formula: CdS
Melting Pt: 980 °C (subl.)
Density: 4,82
Storage Temperature: Ambient
MDL Number: MFCD00010922
CAS Number: 1306-23-6
EINECS: 215-147-8
UN: 3077
ADR: 9,III
Merck Index: 14,1628

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR