Zinc sulphide, Patinal®
Supplier: Merck
Synonyms:
Zinc (II) sulphide
1.08898.0100
1.08898.1000
1.08898.0100EA
0
EUR
1.08898.0100
1.08898.1000
Zinc sulphide, Patinal®
Zinc sulphide
Formula:
SZn MW: 97,46 g/mol Melting Pt: 1064 °C Density: 4,1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011301 CAS Number: 1314-98-3 EINECS: 215-251-3 Merck Index: 12,10294 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...