Yttrium(III) fluoride, Patinal®
Supplier: Merck
Synonyms:
Yttrium trifluoride, Yttrium(III) fluoride
M-Clarity™ quality level = MQ200
1.08707.0100
1.08707.1000
1.08707.0100EA
0
EUR
1.08707.0100
1.08707.1000
Yttrium(III) fluoride, Patinal®
Yttrium (III) fluoride
Formula:
F₃Y MW: 145,9 g/mol Boiling Pt: 2227 °C (1013 hPa) Melting Pt: 1387 °C Density: 4,01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049714 CAS Number: 13709-49-4 EINECS: 237-257-5 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...