Yttrium(III) iodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
|
Warning
|
ampouled under argon
35744.03
35744.06
35744.03EA
176
EUR
35744.03
35744.06
Yttrium(III) iodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Yttrium (III) iodide
Purity: 99.9%
CAS No.: 13470-38-7
Molecular Formula: YI3
Formula Weight: 469.62
Physical Form: Powder/Chunks, Ampouled under argon
Sensitivity: Hygroscopic
MDL No.: MFCD00054144
Formula:
I₃Y MW: 469,62 g/mol Melting Pt: 1004 °C Storage Temperature: Ambient |
MDL Number:
MFCD00054144 CAS Number: 13470-38-7 EINECS: 236-737-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...