Diyttrium trioxide, Patinal®
Supplier: Merck
Synonyms:
Yttrium oxide, Yttrium (III) oxide
1.16440.0100
1.16440.1000
1.16440.0100EA
0
EUR
1.16440.0100
1.16440.1000
Diyttrium trioxide, Patinal®
Diyttrium trioxide
Formula:
Y₂O₃ MW: 225,81 g/mol Boiling Pt: 4300 °C (1013 hPa) Melting Pt: 2410 °C Density: 5,01 g/cm³ (20 °C) |
MDL Number:
MFCD00011473 CAS Number: 1314-36-9 EINECS: 215-233-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...