Titanium(IV) fluoride ≥98%
Supplier: Thermo Scientific
|
Danger
|
11540.14
11540.22
11540.14EA
141
EUR
11540.14
11540.22
Titanium(IV) fluoride ≥98%
Titanium (IV) fluoride
Purity: 98%
CAS No.: 7783-63-3
Molecular Formula: TiF4
Formula Weight: 123.89
Physical Form: Crystalline
Density: 2.798
Appearance: White
Odour: Odourless
Melting Point: 284° subl.
Merck Reference: 14,9479
UN No.: 3260
MDL No.: MFCD00042545
Formula:
F₄Ti MW: 123,87 g/mol Boiling Pt: 284 °C (1013 hPa) Melting Pt: 284 °C (subl.) Density: 2,798 Storage Temperature: Ambient |
MDL Number:
MFCD00042545 CAS Number: 7783-63-3 EINECS: 232-017-6 UN: 3260 ADR: 8,II Merck Index: 14,9479 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...