Hafnium dioxide, Patinal®
Supplier: Merck
1.12450.0100
1.12450.1000
1.12450.0100EA
409
EUR
1.12450.0100
1.12450.1000
Hafnium dioxide, Patinal®
Hafnium dioxide
Formula:
HfO₂ MW: 210,49 g/mol Boiling Pt: 5400 °C (1013 hPa) Melting Pt: 2812 °C Density: 9,68 g/cm³ (20 °C) |
MDL Number:
MFCD00003565 CAS Number: 12055-23-1 EINECS: 235-013-2 Merck Index: 14,04727 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...