Titanium(IV) oxide, Patinal®
Supplier: Merck
Synonyms:
Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide
1.11771.0100
1.11771.1000
1.11771.0100EA
0
EUR
1.11771.0100
1.11771.1000
Titanium(IV) oxide, Patinal®
Titanium (IV) oxide
Formula:
TiO₂ MW: 79,88 g/mol Boiling Pt: 2900 °C (1013 hPa) Melting Pt: 1855 °C Density: 4,2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011269 CAS Number: 13463-67-7 EINECS: 236-675-5 Merck Index: 13,09549 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...