Tantalum(V) oxide 99.99% (trace metals basis)

Supplier: Thermo Scientific

Synonyms: Tantalum(V) oxide, Ditantalum pentaoxide

194170100 194170500
ACRO194170100EA 70.4 EUR
ACRO194170100 ACRO194170500
Tantalum(V) oxide 99.99% (trace metals basis)
Tantalum (V) oxide
Formula: O₅Ta₂
MW: 441,89 g/mol
Melting Pt: 1872 °C
Density: 8,2 g/cm³ (20 °C)
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2
Merck Index: 14,09056

Order Now

Specification Test Results

Appearance White to light beige Powder
Total trace metal impurities ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR