Tantalum(V) oxide, Patinal®
1.08172.1000
1.08172.0100
1.08172.1000EA
0
EUR
1.08172.1000
1.08172.0100
Tantalum(V) oxide, Patinal®
Tantalum (V) oxide
Formula:
O₅Ta₂ MW: 441,89 g/mol Melting Pt: 1870…1880 °C Density: 8,2 g/cm³ (20 °C) |
MDL Number:
MFCD00011254 CAS Number: 1314-61-0 EINECS: 215-238-2 Merck Index: 14,09056 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...