Manganese(II) oxide ≥99.99% (metals basis)

Supplier: Thermo Fisher Scientific
Warning

44441.09 44441.18
44441.09EA 184 EUR
44441.09 44441.18
Manganese(II) oxide ≥99.99% (metals basis)
Manganese (II) oxide

Purity: 99.99% CAS No.: 1344-43-0 Molecular Formula: MnO Formula Weight: 70.94 Physical Form: Powder/Chunks Density: 5.0 Appearance: Green Odour: Odourless MDL No.: MFCD00016226

Formula: MnO
Melting Pt: 1840 °C
Density: 5,0
Storage Temperature: Ambient
MDL Number: MFCD00016226
CAS Number: 1344-43-0
EINECS: 215-695-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR