Platinum(II) chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Platinum dichloride

A16549.03 A16549.06
A16549.03EA 227 EUR
A16549.03 A16549.06
Platinum(II) chloride ≥98%
Platinum (II) chloride

Purity: 98% CAS No.: 10025-65-7 Molecular Formula: Cl2Pt Formula Weight: 266.00 Melting Point: 581° dec. Merck Reference: 14,7528 MDL No.: MFCD00011181

Formula: Cl₂Pt
Melting Pt: 581 °C (dec.)
Density: 5,87
Storage Temperature: Ambient
MDL Number: MFCD00011181
CAS Number: 10025-65-7
EINECS: 233-034-1
UN: 3260
ADR: 8,III
Merck Index: 14,7528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR