Iridium(IV) iodide ≥99.95% (metals basis) Ir ≥ 27.0%, Premion®

Supplier: Thermo Scientific
Warning

40424.03 40424.06
40424.03EA 245 EUR
40424.03 40424.06
Iridium(IV) iodide ≥99.95% (metals basis) Ir ≥ 27.0%, Premion®
Iridium (IV) iodide

Purity: 99.95% CAS No.: 7790-45-6 Molecular Formula: IrI4 Formula Weight: 699.82 Physical Form: Powder Solubility: Insoluble in water and alcohol. Soluble in aqueous KI MDL No.: MFCD00049960

Formula: I₄Ir
MW: 699,84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049960
CAS Number: 7790-45-6
EINECS: 232-206-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR