(±)-γ-Decalactone ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Hexyldihydrofuran-2(3H)-one, Gamma-decalactone, (±)-Decan-4-olide

A19732.06 A19732.14 A19732.22
A19732.06EA 30.5 EUR
A19732.06 A19732.14 A19732.22
(±)-γ-Decalactone ≥98%
(±)-γ-Decalactone

Purity: 98% CAS No.: 706-14-9 Molecular Formula: C10H18O2 Formula Weight: 170.25 Flash Point: 146°(294°F) Refractive Index: 1.500 Boiling Point: 150-153°/15mm Beilstein Reference: 117547 MDL No.: MFCD00005404

Formula: C₁₀H₁₈O₂
MW: 170,25 g/mol
Boiling Pt: 150…153 °C (15 mmHg)
Density: 0,948
Flash Pt: 146 °C (294 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005404
CAS Number: 706-14-9
EINECS: 211-892-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR