2-Chloropyrazine ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Chloropyrazine
A10108.06
A10108.09
A10108.14
A10108.22
A10108.06EA
61.5
EUR
A10108.06
A10108.09
A10108.14
A10108.22
2-Chloropyrazine ≥98%
2-Chloropyrazine
Purity: 98%
CAS No.: 14508-49-7
Molecular Formula: C4H3ClN2
Formula Weight: 114.54
Flash Point: 57°(134°F)
Refractive Index: 1.5370
Boiling Point: 153-154°
Beilstein Reference: 107019
UN No.: 1993
MDL No.: MFCD00006124
Formula:
C₄H₃ClN₂ MW: 114,53 g/mol Boiling Pt: 153…154 °C Density: 1,284 Flash Pt: 57 °C (134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006124 CAS Number: 14508-49-7 EINECS: 238-517-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...