1-Methyl-1H-pyrazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Methylpyrazole
L14831.14
L14831.06
L14831.03
L14831.14EA
206
EUR
L14831.14
L14831.06
L14831.03
1-Methyl-1H-pyrazole ≥97%
1-Methyl-1H-pyrazole
Purity: 97+%
CAS No.: 930-36-9
Molecular Formula: C4H6N2
Formula Weight: 82.11
Flash Point: 35°(95°F)
Refractive Index: 1.4760
Odour: Pungent
Boiling Point: 127°
Beilstein Reference: 105207
UN No.: 1993
MDL No.: MFCD00144943
Formula:
C₄H₆N₂ MW: 82,11 g/mol Boiling Pt: 127 °C Density: 0,988 Flash Pt: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00144943 CAS Number: 930-36-9 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...