4-Bromo-2-chloropyridine ≥94%
Supplier: Thermo Fisher Scientific
|
Danger
|
H27299.14
H27299.06
H27299.14EA
124
EUR
H27299.14
H27299.06
4-Bromo-2-chloropyridine ≥94%
4-Bromo-2-chloropyridine
Purity: 94%
CAS No.: 73583-37-6
Molecular Formula: C5H3BrClN
Formula Weight: 192.44
Flash Point: 104°(219°F)
Melting Point: 18-20°
Boiling Point: 70°/3mm
MDL No.: MFCD03840756
Formula:
C₅H₃BrClN MW: 192.44 g/mol Boiling Pt: 70 °C (3 mmHg) Melting Pt: 18…20 °C Density: 1.734 Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03840756 CAS Number: 73583-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...