2-Amino-5-bromo-3-iodopyridine 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Bromo-3-iodo-2-pyridylamine, 5-Bromo-3-iodopyridin-2-amine
H50002.06
H50002.06EA
477
EUR
H50002.06
2-Amino-5-bromo-3-iodopyridine 98%
2-Amino-5-bromo-3-iodopyridine
Purity: 98%
CAS No.: 381233-96-1
Molecular Formula: C5H4BrIN2
Formula Weight: 298.91
Melting Point: 113-115°
MDL No.: MFCD06659000
Formula:
C₅H₄BrIN₂ MW: 298,91 g/mol Melting Pt: 113…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659000 CAS Number: 381233-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...