2-Amino-6-bromopyridine 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-Bromo-2-pyridylamine, 6-Bromopyridin-2-amine

376870050 376870250 376871000
ACRO376870050EA 81.7 EUR
ACRO376870050 ACRO376870250 ACRO376871000
2-Amino-6-bromopyridine 98%
2-Amino-6-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00137843
CAS Number: 19798-81-3
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Light yellow to light brown Powder
Infrared spectrum Conforms
Melting point 86 to 90 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR