2-Amino-5-bromo-3-nitropyridine 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
5-Bromo-3-nitropyridin-2-amine, 5-Bromo-3-nitro-2-pyridylamine
A14985.14
A14985.06
A14985.14EA
138
EUR
A14985.14
A14985.06
2-Amino-5-bromo-3-nitropyridine 97%
2-Amino-5-bromo-3-nitropyridine
Purity: 97%
CAS No.: 6945-68-2
Molecular Formula: C5H4BrN3O2
Formula Weight: 218.01
Melting Point: 206-210°
Beilstein Reference: 383851
Formula:
C₅H₄BrN₃O₂ MW: 218.01 g/mol Melting Pt: 206…210 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047441 CAS Number: 6945-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...