3-Bromo-2-ethoxypyridine 95%

Supplier: Thermo Scientific
Danger

H27690.MD H27690.03
H27690.MDEA 84 EUR
H27690.MD H27690.03
3-Bromo-2-ethoxypyridine 95%
3-Bromo-2-ethoxypyridine

Purity: 95% CAS No.: 57883-25-7 Molecular Formula: C7H8BrNO Formula Weight: 202.05 MDL No.: MFCD00234309

Formula: C₇H₈BrNO
MW: 202,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234309
CAS Number: 57883-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR