3,4,5-Trichloropyridine ≥98%
Supplier: Thermo Scientific
|
Warning
|
A12587.03
A12587.06
A12587.03EA
50.8
EUR
A12587.03
A12587.06
3,4,5-Trichloropyridine ≥98%
3,4,5-Trichloropyridine
Purity: 98%
CAS No.: 33216-52-3
Molecular Formula: C5H2Cl3N
Formula Weight: 182.44
Melting Point: 70-74°
Boiling Point: 213-215°
MDL No.: MFCD00051685
Formula:
C₅H₂Cl₃N MW: 182,44 g/mol Boiling Pt: 213…215 °C Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051685 CAS Number: 33216-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...