6-Chloronicotinic acid 99%

Supplier: Thermo Fisher Scientific
Warning

168690050 168690250 168691000
ACRO168690050EA 43.7 EUR
ACRO168690050 ACRO168690250 ACRO168691000
6-Chloronicotinic acid 99%
6-Chloronicotinic acid
Formula: C₆H₄ClNO₂
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00006241
CAS Number: 5326-23-8
EINECS: 226-201-5

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 186 to 190 °C
Water ≤0.2 % (K.F.)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR