3,5-Dichloro-2-fluoropyridine ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Fluoro-3,5-dichloropyridine

L20039.03 L20039.14
L20039.03EA 0 EUR
L20039.03 L20039.14
3,5-Dichloro-2-fluoropyridine ≥98%
3,5-Dichloro-2-fluoropyridine

Purity: 98% CAS No.: 823-56-3 Molecular Formula: C5H2Cl2FN Formula Weight: 165.98 Melting Point: 41-43° Beilstein Reference: 1365267

Formula: C₅H₂Cl₂FN
MW: 165,98 g/mol
Melting Pt: 41…43 °C
Storage Temperature: Ambient
MDL Number: MFCD04039349
CAS Number: 823-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR