2-Chloro-3-iodopyridine ≥97%

Supplier: Thermo Scientific
Danger

L20028.06
L20028.06EA 67.2 EUR
L20028.06
2-Chloro-3-iodopyridine ≥97%
2-Chloro-3-iodopyridine

Purity: 97% CAS No.: 78607-36-0 Molecular Formula: C5H3ClIN Formula Weight: 239.44 Melting Point: 93-97° Beilstein Reference: 4243304

Formula: C₅H₃ClIN
MW: 239,44 g/mol
Melting Pt: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00661298
CAS Number: 78607-36-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR